Skip to Content
Merck
All Photos(1)

Key Documents

700746

Sigma-Aldrich

(S)-SDP

≥95%

Synonym(s):

(S)-(−)-7,7′-Bis(diphenylphosphino)-2,2′,3,3′-tetrahydro-1,1′-spirobiindene, (S)-7,7′-Bis(diphenylphosphino)-1,1′-spirobiindane

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$319.00

$319.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$319.00

About This Item

Empirical Formula (Hill Notation):
C41H34P2
CAS Number:
Molecular Weight:
588.66
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$319.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

solid

optical activity

[α]22/D −191°, c = 1 in chloroform

mp

199-204 °C

functional group

phosphine

storage temp.

−20°C

SMILES string

C1CC2(CCc3cccc(P(c4ccccc4)c5ccccc5)c23)c6c1cccc6P(c7ccccc7)c8ccccc8

InChI

1S/C41H34P2/c1-5-17-33(18-6-1)42(34-19-7-2-8-20-34)37-25-13-15-31-27-29-41(39(31)37)30-28-32-16-14-26-38(40(32)41)43(35-21-9-3-10-22-35)36-23-11-4-12-24-36/h1-26H,27-30H2

InChI key

DKZNVWNOOLQCMJ-UHFFFAOYSA-N

General description

(S)-SDP is a chiral diphosphine reagent used in the asymmetric hydrogenation of ketones catalyzed by transition metal. It is also used in hydrosilylation or cyclization of 1,6-enynes.[1]

Application

(S)-SDP can be used:
  • As a ligand in the ruthenium-catalyzed asymmetric hydrogenation of cyclic ketones to give cycloalkanols.[2]
  • To prepare ruthenium complexes, which are used in the asymmetric hydrogenation of different prochiral ketones.[3]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic Kinetic Resolution Allows a Highly Enantioselective Synthesis of cis-alpha-Aminocycloalkanols by Ruthenium-Catalyzed Asymmetric Hydrogenation
Liu S, et al.
Angewandte Chemie (International ed. in English), 46(39), 7506-7508 (2007)
Synthesis of spiro diphosphines and their application in asymmetric hydrogenation of ketones
Xie J, et al.
Journal of the American Chemical Society, 125(15), 4404-4405 (2003)
(R?and (S)?7,7′?Bis(diphenylphosphino)?2,2′,3,3′?tetrahydro?1,1′?spirobiindene
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service