Skip to Content
Merck
All Photos(1)

Key Documents

69723

Sigma-Aldrich

7-Methylxanthine

≥98.0% (HPLC)

Synonym(s):

2,6-Dihydroxy-7-methylpurine, Heteroxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14
Beilstein:
171027
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

form

powder

mp

≥300 °C

SMILES string

Cn1cnc2NC(=O)NC(=O)c12

InChI

1S/C6H6N4O2/c1-10-2-7-4-3(10)5(11)9-6(12)8-4/h2H,1H3,(H2,8,9,11,12)

InChI key

PFWLFWPASULGAN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

7-Methylxanthine is an oxopurine, which belongs to the class of xanthines. It may be synthesized by the reaction between 4-amino-1-methylimidazole-5-carboxamide and diethyl carbonate.[1][2]

Application

7-Methylxanthine can be used as a building block to synthesize tricyclic imidazo[2,1-i]purinone derivatives as potential adenosine receptor antagonists.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Trier et al.
The British journal of ophthalmology, 83(12), 1370-1375 (1999-11-27)
To examine a possible effect of 7-methylxanthine, theobromine, acetazolamide, or L-ornithine on the ultrastructure and biochemical composition of rabbit sclera. Groups of pigmented rabbits, six in each group, were dosed during 10 weeks with one of the substances under investigation
Swati Sucharita Dash et al.
Current microbiology, 55(1), 56-60 (2007-06-08)
In this study, the kinetics of degradation of caffeine and related methylxanthines by induced cells of Pseudomonas sp. was performed. The kinetics data showed that degradation of caffeine, theobromine, and 7-methylxanthine followed Michealis-Menten kinetics. The values of K (m) are
Dongmei Cui et al.
Acta ophthalmologica, 89(4), 328-334 (2009-10-29)
The aim of this study was to determine the effect of the adenosine receptor antagonist 7-methylxanthine (7-MX) on form deprivation myopia in 3-week-old guinea pigs. Two groups of 3-week-old guinea pigs were subjected to monocular deprivation (MD) using a diffuser
Kouichi Mizuno et al.
FEBS letters, 534(1-3), 75-81 (2003-01-16)
In coffee and tea plants, caffeine is synthesized from xanthosine via a pathway that includes three methylation steps. We report the isolation of a bifunctional coffee caffeine synthase (CCS1) clone from coffee endosperm by reverse transcription-polymerase chain reaction (RT-PCR) and
H M Himmel et al.
Journal of chromatography, 568(1), 105-115 (1991-07-17)
A high-performance liquid chromatographic method is described for the separation and quantitation of several purine compounds, including hypoxanthine. The isocratic separation of a standard mixture of nine compounds is achieved within 20 min on a reversed-phase Nucleosil 100-5C18 column, with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service