Skip to Content
Merck
All Photos(1)

Key Documents

69701

Sigma-Aldrich

1-Methyl-2-vinylpyridinium triflate

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10F3NO3S
CAS Number:
Molecular Weight:
269.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (T)

form

solid

SMILES string

[O-]S(=O)(=O)C(F)(F)F.C[n+]1ccccc1C=C

InChI

1S/C8H10N.CHF3O3S/c1-3-8-6-4-5-7-9(8)2;2-1(3,4)8(5,6)7/h3-7H,1H2,2H3;(H,5,6,7)/q+1;/p-1

InChI key

VJUSJWUMYXKNPL-UHFFFAOYSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diederik S Laman Trip et al.
Nature microbiology, 5(7), 943-954 (2020-04-22)
The conventional view is that high temperatures cause microorganisms to replicate slowly or die. In this view, microorganisms autonomously combat heat-induced damages. However, microorganisms co-exist with each other, which raises the underexplored and timely question of whether microorganisms can cooperatively
Mirella Kanerva et al.
The Science of the total environment, 742, 140259-140259 (2020-07-30)
The fitness and recruitment of fish stocks can be markedly affected by environmental disturbances including global warming, eutrophication and contamination. Understanding the effects of environmental stressors on salmon physiology during marine residence is of a global concern as marine survival
Nuntiya Somparn et al.
Free radical research, 53(7), 791-799 (2019-06-15)
Oxidative stress caused as a result of iron overload is implicated in clinical manifestation of beta-thalassemia/haemoglobin E (β-Thal/HbE). In this study, we investigated the cellular adaptation against oxidative stress in β-Thal/HbE patients. Twenty-four paediatric β-Thal/HbE patients and 22 healthy controls
Jaroon Wandee et al.
Life sciences, 217, 155-163 (2018-12-12)
Metformin (Met), an essential antidiabetic agent, shows antitumor activity in some cancers. A previous study showed that Met enhanced cytotoxic activity of cisplatin (Cis) in cholangiocarcinoma (CCA) in association with the activation of AMP-activated protein kinase and suppression of Akt-mTOR.
Ornanong Tusskorn et al.
Naunyn-Schmiedeberg's archives of pharmacology, 391(6), 657-667 (2018-04-19)
Phenethyl isothiocyanate (PEITC) is a potential cancer prevention agent that is found in cruciferous vegetables. Previous studies have shown that the effect of PEITC-induced cell death declines rapidly after administration. The metabolic fate of PEITC is modulated by glutathione S-transferases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service