Skip to Content
Merck
All Photos(1)

Documents

696420

Sigma-Aldrich

(R)-Ethyl piperidine-3-carboxylate

97%

Synonym(s):

(R)-Piperidine-3-carboxylic acid ethyl ester, (R)-ethyl nipecotate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO2
CAS Number:
Molecular Weight:
157.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

density

1.092 g/mL at 25 °C

SMILES string

CCOC(=O)[C@@H]1CCCNC1

InChI

1S/C8H15NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h7,9H,2-6H2,1H3/t7-/m1/s1

InChI key

XIWBSOUNZWSFKU-SSDOTTSWSA-N

Application

Reactant for synthesis of:
  • DPP-4 inhibitors
  • Dual H1/5-HT2A receptor antagonists for treatment of sleep disorders
  • Serotonin and noradrenaline reuptake inhibitors
  • GABA uptake inhibitors

Reactant for stereocontrolled hydoiodination of alkynes

Used for an orally potent anti-anxiety drug

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F

Flash Point(C)

104 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S H Zorn et al.
The Journal of pharmacology and experimental therapeutics, 242(1), 173-178 (1987-07-01)
Previous reports have suggested that the ethyl ester of (R)-nipecotic acid ethyl ester [(R)-NAEE] displays cholinomimetic properties in vivo. The present study was undertaken to characterize more fully this action by examining the effects of (R)-NAEE in a number of
T Andree et al.
Life sciences, 32(19), 2265-2272 (1983-05-09)
THIP (4,5,6,7-tetrahydroisoxazolo (5,4-c) pyridone-3-ol), a direct acting GABA receptor agonist, has been shown to have antinociceptive properties. To determine whether tolerance develops to the analgesic response, mice received multiple injections of THIP for up to 21 days after which analgesia
M Potegal
Psychopharmacology, 89(4), 444-448 (1986-01-01)
The GABA uptake inhibitor ethyl (R,S)-nipecotate produces a dose-dependent suppression of aggression in highly aggressive hamsters but not in minimally aggressive ones. This suppression occurs at doses below those producing peripheral cholinergic effects; at the highest dose used it persists
Wen-Juan Zhao et al.
Acta pharmacologica Sinica, 24(10), 991-995 (2003-10-09)
The changes of seizure susceptibility of transgenic mice overexpressing GABA transporter-1 (GAT-1) were studied to clarify the possible role of GABAergic transmission in epileptogenesis. Seizures were induced by intraperitoneal administration of pentylenetetrazol (PTZ), picrotoxin (PIC), or kainic acid (KA) respectively.
A M Rustum et al.
Journal of pharmaceutical and biomedical analysis, 17(8), 1439-1447 (1998-11-04)
Chiral HPLC methods were developed and validated for tiagabine.HCl and its two chiral precursors to determine the chiral purity of the three compounds to ensure the quality of the final product which is used as a new antiepileptic drug. Tiagabine.HCl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service