Skip to Content
Merck
All Photos(1)

Key Documents

695157

Sigma-Aldrich

Bis(3,5-dimethylphenyl)phosphine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$323.00

$323.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$323.00

About This Item

Empirical Formula (Hill Notation):
C16H19P
CAS Number:
Molecular Weight:
242.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$323.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.599

density

1.017 g/mL at 25 °C

functional group

phosphine

SMILES string

Cc1cc(C)cc(Pc2cc(C)cc(C)c2)c1

InChI

1S/C16H19P/c1-11-5-12(2)8-15(7-11)17-16-9-13(3)6-14(4)10-16/h5-10,17H,1-4H3

InChI key

GPFIUEZTNRNFGD-UHFFFAOYSA-N

Application

Bis(3,5-dimethylphenyl)phosphine can be used as a reactant to prepare:
  • Iron(II) chiral diimine diphosphine complexes as catalysts for the asymmetric transfer hydrogenation of ketones.[1]
  • The complex of iridium with biphenol phosphite-phosphine bidentate ligand as an asymmetric catalyst for the hydrogenation of N-arylimines.[2]
  • Chiral phosphines with excellent stereoselectivity by 1, 4 addition reaction with α,β-unsaturated aldehydes using a palladium catalyst.[3]

Legal Information

Product of Kanata Chemical Technologies, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iridium Complexes with Phosphine- Phosphite Ligands. Structural Aspects and Application in the Catalytic Asymmetric Hydrogenation of N-Aryl Imines
Vargas S, et al.
Organometallics, 25(4), 961-973 (2006)
Stereoelectronic factors in iron catalysis: synthesis and characterization of aryl-substituted iron (II) carbonyl P-N-N-P complexes and their use in the asymmetric transfer hydrogenation of ketones
Sues PE, et al.
Organometallics, 30(16), 4418-4431 (2011)
Palladium-catalyzed 1, 4-addition of diarylphosphines to α, β -unsaturated aldehydes
Chen Y-R and Duan W-L
Organic Letters, 13(21), 5824-5826 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service