Skip to Content
Merck
All Photos(1)

Key Documents

69418

Sigma-Aldrich

(S)-Boc-3-(trifluoromethyl)-β-Phe-OH

≥98.0%

Synonym(s):

(S)-3-(Boc-amino)-3-[3-(trifluoromethyl)phenyl]propionic acid, Boc-3-(trifluoromethyl)-D-β-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18F3NO4
CAS Number:
Molecular Weight:
333.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0%

form

powder

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CC(O)=O)c1cccc(c1)C(F)(F)F

InChI

1S/C15H18F3NO4/c1-14(2,3)23-13(22)19-11(8-12(20)21)9-5-4-6-10(7-9)15(16,17)18/h4-7,11H,8H2,1-3H3,(H,19,22)(H,20,21)/t11-/m0/s1

InChI key

UPKLBXIBSSZUID-NSHDSACASA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abhilash Padavannil et al.
Bio-protocol, 10(10), e3625-e3625 (2021-03-05)
Histones constitute the protein components of nucleosomes. Despite their small sizes, histones do not diffuse through the nuclear pore complex. Instead, they are transported to the nucleus by importins, either alone or in complex with histone chaperones. Determining the molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service