Skip to Content
Merck
All Photos(1)

Key Documents

692964

Sigma-Aldrich

(S)-RuCl[(p-cymene)(T-BINAP)]Cl

Synonym(s):

Chloro[(S)-(−)-2,2′-bis(di-p-tolylphosphino)-1,1′-binaphthyl](p-cymene)ruthenium(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C58H54ClP2Ru]+Cl
CAS Number:
Molecular Weight:
984.97
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

powder

storage temp.

2-8°C

SMILES string

[Cl-].Cl[Ru+].C[C]1[CH][CH][C]([CH][CH]1)C(C)C.Cc2ccc(cc2)P(c3ccc(C)cc3)c4ccc5ccccc5c4-c6c(ccc7ccccc67)P(c8ccc(C)cc8)c9ccc(C)cc9

InChI

1S/C48H40P2.C10H14.2ClH.Ru/c1-33-13-23-39(24-14-33)49(40-25-15-34(2)16-26-40)45-31-21-37-9-5-7-11-43(37)47(45)48-44-12-8-6-10-38(44)22-32-46(48)50(41-27-17-35(3)18-28-41)42-29-19-36(4)20-30-42;1-8(2)10-6-4-9(3)5-7-10;;;/h5-32H,1-4H3;4-8H,1-3H3;2*1H;/q;;;;+2/p-2

InChI key

BHIUOOHVIWKUTN-UHFFFAOYSA-L

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogenation, Asymmetric Catalysis, Binap, SEGPHOS®, Aldol reaction, Alkenylation, Arylation, Mannich reaction, Fluorination, Michael addition, Hydrosilylation, Cycloaddition, Takasago

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service