About This Item
Assay
98%
form
powder
reaction suitability
reagent type: catalyst
core: palladium
SMILES string
[Cu]I.[Cu]I.Cl[Pd]Cl.Cl[Pd]Cl.Cl[Pd]Cl.Cl[Pd]Cl.Cl[Pd]Cl.Clc1cccnc1.Clc2cccnc2.Clc3cccnc3.Clc4cccnc4.Clc5cccnc5.CC(C)c6cccc(C(C)C)c6N7CN(C=C7)c8c(cccc8C(C)C)C(C)C.CC(C)c9cccc(C(C)C)c9N%10CN(C=C%10)c%11c(cccc%11C(C)C)C(C)C.CC(C)c%12cccc(C(C)C)c%12N%13CN(C=C%13)c%14c(cccc%14C(C)C)C(C)C.CC(C)c%15cccc(C(C)C)c%15N%16CN(C=C%16)c%17c(cccc%17C(C)C)C(C)C.CC(C)c%18cccc(C(C)C)c%18N%19CN(C=C%19)c%20c(cccc%20C(C)C)C(C)C
InChI
1S/5C27H38N2.5C5H4ClN.10ClH.2Cu.2HI.5Pd/c5*1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;5*6-5-2-1-3-7-4-5;;;;;;;;;;;;;;;;;;;/h5*9-16,18-21H,17H2,1-8H3;5*1-4H;10*1H;;;2*1H;;;;;/q;;;;;;;;;;;;;;;;;;;;2*+1;;;5*+2/p-12
InChI key
HHZQXZQIQVCMBL-UHFFFAOYSA-B
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral
Target Organs
Thyroid
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service