Skip to Content
Merck
All Photos(1)

Documents

691445

Sigma-Aldrich

4-Amino-3-chlorobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClNO2
CAS Number:
Molecular Weight:
171.58
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

224-229 °C

application(s)

peptide synthesis

SMILES string

Nc1ccc(cc1Cl)C(O)=O

InChI

1S/C7H6ClNO2/c8-5-3-4(7(10)11)1-2-6(5)9/h1-3H,9H2,(H,10,11)

InChI key

YIYBPEDZAUFQLO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z W She et al.
Free radical biology & medicine, 22(6), 989-998 (1997-01-01)
This study was designed to develop traps for hypochlorous acid (HOCl) which could be used to detect HOCl in the microenvironment of activated neutrophils. Reagent HOCl was found to react with para-aminobenzoic acid (PABA) in aqueous solution to produce a
Tomasz Frączek et al.
Journal of enzyme inhibition and medicinal chemistry, 31(3), 481-489 (2015-05-06)
Azoles are a promising class of the new generation of HIV-1 non-nucleoside reverse transcriptase inhibitors (NNRTIs). From thousands of reported compounds, many possess the same basic structure of an aryl substituted azole ring linked by a thioglycolamide chain with another

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service