Skip to Content
Merck
All Photos(1)

Key Documents

691437

Sigma-Aldrich

4-Amino-2-methylbenzoic acid

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$107.00

$107.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$107.00

About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$107.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

160-165 °C

application(s)

peptide synthesis

SMILES string

Cc1cc(N)ccc1C(O)=O

InChI

1S/C8H9NO2/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4H,9H2,1H3,(H,10,11)

InChI key

XRSQZFJLEPBPOZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weixiang Liu et al.
Carbohydrate polymers, 160, 97-105 (2017-01-25)
A novel type of O-carboxymethyl chitosan Schiff bases (O-CSPX) was synthesized via a condensation reaction. After the coordination reaction of cupric ions, Cu(II) complexes (O-CSPX-Cu) were achieved. The theoretical structure of O-CSPX-Cu calculated by Gaussian 09 reveals that the copper

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service