Skip to Content
Merck
All Photos(1)

Key Documents

69066

Sigma-Aldrich

Calix[8]arene

technical, ≥90% (CH)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$129.00

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$129.00

About This Item

Empirical Formula (Hill Notation):
C56H48O8
CAS Number:
Molecular Weight:
848.98
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical

Quality Level

Assay

≥90% (CH)

form

powder

SMILES string

Oc1c2Cc3cccc(Cc4cccc(Cc5cccc(Cc6cccc(Cc7cccc(Cc8cccc(Cc9cccc(Cc1ccc2)c9O)c8O)c7O)c6O)c5O)c4O)c3O

InChI

1S/C56H48O8/c57-49-33-9-1-10-34(49)26-36-12-3-14-38(51(36)59)28-40-16-5-18-42(53(40)61)30-44-20-7-22-46(55(44)63)32-48-24-8-23-47(56(48)64)31-45-21-6-19-43(54(45)62)29-41-17-4-15-39(52(41)60)27-37-13-2-11-35(25-33)50(37)58/h1-24,57-64H,25-32H2

InChI key

HDPRHRZFFPXZIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Calix[8]arene can be used:
  • To prepare water-soluble glycocalix[8]arenes, which are used as model compounds to study the carbohydrate-protein binding interactions.[1]
  • As a starting material in the synthesis of calix[8]arene sulfonic acids of biological importance.[2]
  • As a ligand in the synthesis of tetranuclear calix[8]arene bismuth complexes.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis of bismuth and antimony complexes of the ?larger? calix [n] arenes (n= 6--8); from mononuclear to tetranuclear complexes
Mendoza-Espinosa D, et al.
Dalton Transactions, 16(26), 5226-5238 (2009)
Calix[8] arene-based glycoconjugates as multivalent carbohydrate-presenting systems
Consoli GML, et al.
Tetrahedron Letters, 44(40), 7467-7470 (2003)
Bence György et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 25(2), 379-391 (2017-01-14)
Adeno-associated virus (AAV) is a safe and effective vector for gene therapy for retinal disorders. Gene therapy for hearing disorders is not as advanced, in part because gene delivery to sensory hair cells of the inner ear is inefficient. Although
Design, synthesis and anticoagulant activity of new flexible calix [8] arene sulfonic acids
Rekkab S, et al.
Comptes Rendus Chimie, 16(7), 672-678 (2013)
Kamilla M E Laidlaw et al.
Journal of cell science, 134(2) (2021-01-15)
Eukaryotic cells adapt their metabolism to the extracellular environment. Downregulation of surface cargo proteins in response to nutrient stress reduces the burden of anabolic processes whilst elevating catabolic production in the lysosome. We show that glucose starvation in yeast triggers

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service