Skip to Content
Merck
All Photos(1)

Key Documents

689998

Sigma-Aldrich

O-[2-(Biotinylamino)ethyl]-O′-(2-carboxyethyl)undecaethylene glycol

≥95% (oligomer purity)

Synonym(s):

Biotin-PEG acid (n = 11)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$854.00

$854.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$854.00

About This Item

Empirical Formula (Hill Notation):
C37H69N3O16S
Molecular Weight:
844.02
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$854.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95% (oligomer purity)

form

powder

reaction suitability

reagent type: cross-linking reagent

Ω-end

carboxylic acid

α-end

biotin

storage temp.

−20°C

SMILES string

OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)CCCCC1SCC2NC(=O)NC12

InChI

1S/C37H69N3O16S/c41-34(4-2-1-3-33-36-32(31-57-33)39-37(44)40-36)38-6-8-46-10-12-48-14-16-50-18-20-52-22-24-54-26-28-56-30-29-55-27-25-53-23-21-51-19-17-49-15-13-47-11-9-45-7-5-35(42)43/h32-33,36H,1-31H2,(H,38,41)(H,42,43)(H2,39,40,44)

InChI key

AEMUKWXFLKZKMD-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service