Skip to Content
Merck
All Photos(1)

Documents

689602

Sigma-Aldrich

4-(Triphenylphosphonio)butane-1-sulfonate

99%

Synonym(s):

Triphenyl-(4-sulfo-butyl)-phosphonium inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23O3PS
CAS Number:
Molecular Weight:
398.46
Beilstein:
4018406
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

≥98.5% (HPLC)
99%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

impurities

≤0.5% water

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤100 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

functional group

phosphine

SMILES string

[O-]S(=O)(=O)CCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C22H23O3PS/c23-27(24,25)19-11-10-18-26(20-12-4-1-5-13-20,21-14-6-2-7-15-21)22-16-8-3-9-17-22/h1-9,12-17H,10-11,18-19H2

InChI key

RKQKXPDRWYNUGZ-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Sulfonyl-functionalized ionic liquids
  • Prostaglandins containing a sulfo group

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service