Skip to Content
Merck
All Photos(1)

Documents

68892

Sigma-Aldrich

Methyl 2-(4-Boc-piperazino)-2-(4-pyridyl)acetate

technical, ≥90% (HPLC)

Synonym(s):

2-(4-Boc-piperazino)-2-(4-pyridyl)acetic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25N3O4
CAS Number:
Molecular Weight:
335.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

technical

Assay

≥90% (HPLC)

form

liquid

application(s)

peptide synthesis

SMILES string

COC(=O)C(N1CCN(CC1)C(=O)OC(C)(C)C)c2ccncc2

InChI

1S/C17H25N3O4/c1-17(2,3)24-16(22)20-11-9-19(10-12-20)14(15(21)23-4)13-5-7-18-8-6-13/h5-8,14H,9-12H2,1-4H3

InChI key

JXZYJTGTWULZIZ-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service