Skip to Content
Merck
All Photos(1)

Key Documents

686301

Sigma-Aldrich

(S)-4-Bromo-α-methylbenzyl alcohol

95%

Synonym(s):

(S)-1-(4-Bromophenyl)ethanol, (S)-4′-Bromo-1-phenylethanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$251.00

$251.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$251.00

About This Item

Linear Formula:
BrC6H4CH(CH3)OH
CAS Number:
Molecular Weight:
201.06
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$251.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

optical activity

[α]20/D -38.0°, c = 1 in chloroform

refractive index

n20/D 1.570

density

1.322 g/mL at 25 °C

functional group

bromo
hydroxyl

SMILES string

C[C@H](O)c1ccc(Br)cc1

InChI

1S/C8H9BrO/c1-6(10)7-2-4-8(9)5-3-7/h2-6,10H,1H3/t6-/m0/s1

InChI key

XTDTYSBVMBQIBT-LURJTMIESA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service