Skip to Content
Merck
All Photos(1)

Key Documents

684554

Sigma-Aldrich

2-(tert-Butyldimethylsilyloxy)-6-bromonaphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21BrOSi
Molecular Weight:
337.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

97%

form

solid

mp

61-64 °C

SMILES string

CC(C)(C)[Si](C)(C)Oc1ccc2cc(Br)ccc2c1

InChI

1S/C16H21BrOSi/c1-16(2,3)19(4,5)18-15-9-7-12-10-14(17)8-6-13(12)11-15/h6-11H,1-5H3

InChI key

SSFFBVWGOVGZJB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service