Skip to Content
Merck
All Photos(1)

Key Documents

683701

Sigma-Aldrich

6,7-Dihydro-2-pentafluorophenyl-5H-pyrrolo[2,1-c]-1,2,4-triazolium tetrafluoroborate

97%

Synonym(s):

2-(Pentafluorophenyl)-2,5,6,7-tetrahydropyrrolo[2,1-c][1,2,4]triazol-4-ium tetrafluoroborate 

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$184.00
500 MG
$785.00

$184.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$184.00
500 MG
$785.00

About This Item

Empirical Formula (Hill Notation):
C11H7BF9N3
CAS Number:
Molecular Weight:
362.99
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$184.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

mp

245 °C

SMILES string

F[B-](F)(F)F.Fc1c(F)c(F)c(c(F)c1F)-[n+]2cn3CCCc3n2

InChI

1S/C11H7F5N3.BF4/c12-6-7(13)9(15)11(10(16)8(6)14)19-4-18-3-1-2-5(18)17-19;2-1(3,4)5/h4H,1-3H2;/q+1;-1

InChI key

KIWCIWCCQVTPOY-UHFFFAOYSA-N

Application

This triazolium catalyst is used for intermolecular homodimerization of aryl aldehydes, intramolecular aldehyde-ketone benzoin cyclizations, and intramolecular Stetter reations.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ming He et al.
Journal of the American Chemical Society, 128(26), 8418-8420 (2006-06-29)
Highly enantioselective, N-heterocyclic carbene (NHC)-catalyzed aza-Diels-Alder reactions are described. A novel chiral triazolium salt based on the cis-1,2-aminoindanol platform serves as an efficient precatalyst for the NHC-catalyzed redox generation of enolate dienophiles that undergo LUMOdiene-controlled Diels-Alder reactions with N-sulfonyl-alpha,beta-unsaturated imines

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service