Skip to Content
Merck
All Photos(2)

Key Documents

683116

Sigma-Aldrich

Chiralyst P294

Umicore

Synonym(s):

(Acetylacetonato)(norbornadiene)rhodium(I), Acetylacetonatonorbornadienerhodium(I), Norbornadiene(acetylacetonato)rhodium, (Acetylacetonato)(bicyclo[2.2.1]hepta-2,5-diene)rhodium(I)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$451.00

$451.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$451.00

About This Item

Empirical Formula (Hill Notation):
C12H15O2Rh
CAS Number:
Molecular Weight:
294.15
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$451.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

mp

176-178 °C (lit.)

SMILES string

C1C2C=CC1C=C2.CC(=O)\C=C(\C)O[Rh]

InChI

1S/C7H8.C5H8O2.Rh/c1-2-7-4-3-6(1)5-7;1-4(6)3-5(2)7;/h1-4,6-7H,5H2;3,6H,1-2H3;/q;;+1/p-1/b;4-3-;

InChI key

IIQPRGKBFZGPIB-LWFKIUJUSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Precatalysts for asymmetric catalysis offer batch-to-batch consistency in a range of catalytic reactions, supporting diverse research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service