Skip to Content
Merck
All Photos(1)

Key Documents

683035

Sigma-Aldrich

1-Trifluoroacetyl piperidine

97%

Synonym(s):

2,2,2-Trifluoro-1-(piperidin-1-yl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10F3NO
CAS Number:
Molecular Weight:
181.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4171

density

1.226 g/mL at 25 °C

functional group

fluoro

SMILES string

FC(F)(F)C(=O)N1CCCCC1

InChI

1S/C7H10F3NO/c8-7(9,10)6(12)11-4-2-1-3-5-11/h1-5H2

InChI key

BCJUMGSHTLYECD-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Photoreactive probes that differentiate the binding sites of noncompetitive GABA receptor antagonists
  • Photoactive a-mannosides and mannosyl peptides
  • Inactivators of human cytochrome P450 2B6
  • Photoaffinity labeled fusidic acid analogues
  • Hydroxyamides as anticonvulsants
  • ET-18-OCH3 with antiproliferative properties

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service