Skip to Content
Merck
All Photos(1)

Key Documents

68193

Sigma-Aldrich

Methyl L-3-phenyllactate

≥97.0% (HPLC)

Synonym(s):

Methyl (S)-2-hydroxy-3-phenylpropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O3
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

optical activity

[α]/D +4.4±0.5°, c = 1% in methanol

mp

40-45 °C (lit.)

functional group

ester
hydroxyl
phenyl

SMILES string

COC(=O)[C@@H](O)Cc1ccccc1

InChI

1S/C10H12O3/c1-13-10(12)9(11)7-8-5-3-2-4-6-8/h2-6,9,11H,7H2,1H3/t9-/m0/s1

InChI key

NMPPJJIBQQCOOI-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl L-3-phenyllactate can be used:
  • As an intermediate in the synthesis of bortezomib analogs as 20S proteasome inhibitors.
  • As a substrate in the amination of the benzylic C-H bonds and the aminated products are further used to prepare β-lactams.
  • As an intermediate in one of the key synthetic steps for the preparation of YM-254890 analog, a selective Gαq/11 inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward the selective inhibition of G proteins: total synthesis of a simplified YM-254890 analog
Rensing DT, et al.
Organic Letters, 17(9), 2270-2273 (2015)
Synthesis of Biologically Relevant Compounds by Ruthenium Porphyrin Catalyzed Amination of Benzylic C-H Bonds
Zardi P, et al.
Organometallics, 33(9), 2210-2218 (2014)
Discovery of novel 20S proteasome inhibitors by rational topology-based scaffold hopping of bortezomib
Xu Y, et al.
Bioorganic & Medicinal Chemistry Letters, 28(12), 2148-2152 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service