Skip to Content
Merck
All Photos(1)

Key Documents

681466

Sigma-Aldrich

Ethyl indole-3-carboxylate

96%

Synonym(s):

3-(Ethoxycarbonyl)indole, Indole-3-carboxylic acid ethyl ester, NSC 63796

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

form

solid

mp

120-124 °C

SMILES string

CCOC(=O)c1c[nH]c2ccccc12

InChI

1S/C11H11NO2/c1-2-14-11(13)9-7-12-10-6-4-3-5-8(9)10/h3-7,12H,2H2,1H3

InChI key

XOUHVMVYFOXTMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to 1,2-aziridinopyrrolo-indoles.[1]
Reactant for preparation of:
  • Indole-based heterocycles for synthesis of indole-based p38 inhibitors and gramines
  • Glycine receptor ligands
  • Inhibitors of Human 5-Lipoxygenase
  • Antimicrobial agents

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edwin Vedejs et al.
The Journal of organic chemistry, 69(6), 1794-1799 (2004-04-03)
A convergent synthetic route to the 1,2-aziridinopyrrolo(1,2-a)indole 34 has been developed. Key features of this route include the deuterium kinetic isotope effect to block undesired indole lithiation during tin-lithium exchange from 27a to 30a, the intramolecular Michael addition to generate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service