Skip to Content
Merck
All Photos(1)

Key Documents

680923

Sigma-Aldrich

n-Butylboronic acid pinacol ester

97%

Synonym(s):

2-Butyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21BO2
CAS Number:
Molecular Weight:
184.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n/D 1.4182

density

0.865 g/mL

SMILES string

CCCCB1OC(C)(C)C(C)(C)O1

InChI

1S/C10H21BO2/c1-6-7-8-11-12-9(2,3)10(4,5)13-11/h6-8H2,1-5H3

InChI key

ZHBANXSNVFDSMK-UHFFFAOYSA-N

Application

n-Butylboronic acid pinacol ester can be used to prepare the fusaric acid and its analogs as possible inhibitors of quorum sensing in bacteria.[1]

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Truong Thanh Tung et al.
European journal of medicinal chemistry, 126, 1011-1020 (2016-12-30)
Taking advantage of microwave-assisted synthesis, efficient and expedite procedures for preparation of a library of fusaric acid and 39 analogues are reported. The fusaric acid analogues were tested in cell-based screening assays for inhibition of the las and rhl quorum

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service