Skip to Content
Merck
All Photos(1)

Documents

680559

Sigma-Aldrich

3-Chloro-4-methylphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8BClO2
CAS Number:
Molecular Weight:
170.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

form

solid

mp

228-232 °C

SMILES string

Cc1ccc(cc1Cl)B(O)O

InChI

1S/C7H8BClO2/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4,10-11H,1H3

InChI key

YTJUYWRCAZWVSX-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Application

Reactant involved in:
  • Intromolecular aromatic carbenoid insertion for the synthesis of fluorenes
  • Synthesis of biaryl amides with muscarinic acetylcholine receptor subtype M1 agonistic activity
  • Preparation of borinic acid picolinate esters for use against cutaneous diseases
  • Synthesis of TRPV1 antagonists as treatment for chronic pain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service