Skip to Content
Merck
All Photos(1)

Key Documents

68047

Sigma-Aldrich

4-Borono-D-phenylalanine

≥97.0% (HPLC)

Synonym(s):

4-Dihydroxyboryl-D-phenylalanine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$874.00

$874.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$874.00

About This Item

Empirical Formula (Hill Notation):
C9H12BNO4
CAS Number:
Molecular Weight:
209.01
Beilstein:
9202597
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$874.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Assay

≥97.0% (HPLC)

form

solid

optical activity

[α]/D +6.0±1.0°, c = 0.5 in 2% HCl

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N[C@H](Cc1ccc(cc1)B(O)O)C(O)=O

InChI

1S/C9H12BNO4/c11-8(9(12)13)5-6-1-3-7(4-2-6)10(14)15/h1-4,8,14-15H,5,11H2,(H,12,13)/t8-/m1/s1

InChI key

NFIVJOSXJDORSP-MRVPVSSYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service