Skip to Content
Merck
All Photos(1)

Key Documents

679771

Sigma-Aldrich

Chloro[(1,1′-biphenyl-2-yl)di-tert-butylphosphine]gold(I)

98%

Synonym(s):

(2-Biphenyl)di-tert-butylphosphine gold(I) chloride, 2-(Di-tert-butylphosphino)biphenyl gold(I) chloride, JohnPhos AuCl, [[1,1′-Biphenyl]-2-ylbis(t-butylphosphine]chlorogold

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$397.00

$397.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$397.00

About This Item

Empirical Formula (Hill Notation):
C20H27AuClP
CAS Number:
Molecular Weight:
530.82
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$397.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

237-240 °C

SMILES string

Cl[Au].CC(C)(C)P(c1ccccc1-c2ccccc2)C(C)(C)C

InChI

1S/C20H27P.Au.ClH/c1-19(2,3)21(20(4,5)6)18-15-11-10-14-17(18)16-12-8-7-9-13-16;;/h7-15H,1-6H3;;1H/q;+1;/p-1

InChI key

MZMOUYZCJIFYAH-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Efficient Au(I)-catalyzed intramolecular addition of beta-ketoamide to unactivated alkenes.
Cong-Ying Zhou et al.
Journal of the American Chemical Society, 129(18), 5828-5829 (2007-04-19)

Articles

Access gold precatalysts, silver salts, and unsaturated building blocks to accelerate research success in catalysis applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service