Skip to Content
Merck
All Photos(3)

Key Documents

679674

Sigma-Aldrich

2-Benzyloxy-1-methylpyridinium triflate

96%

Synonym(s):

Bn-OPT, Dudley Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$232.00
5 G
$298.00

$232.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$232.00
5 G
$298.00

About This Item

Empirical Formula (Hill Notation):
C14H14F3NO4S
CAS Number:
Molecular Weight:
349.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$232.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

form

solid

mp

85-91 °C

functional group

fluoro
phenyl
triflate

storage temp.

2-8°C

SMILES string

[O-]S(=O)(=O)C(F)(F)F.C[n+]1ccccc1OCc2ccccc2

InChI

1S/C13H14NO.CHF3O3S/c1-14-10-6-5-9-13(14)15-11-12-7-3-2-4-8-12;2-1(3,4)8(5,6)7/h2-10H,11H2,1H3;(H,5,6,7)/q+1;/p-1

InChI key

DUXHYQYOPHEFGC-UHFFFAOYSA-M

Application

Dudley Reagents


  • Novel reagent for benzyl protection of alcohols under neutral conditions.[1][2] Promotes selective formation of benzyl esters of carboxylic acids in presence of triethylamine.[3]

Legal Information

Sold under license from FSURF, US patent 7,754,909 and related patents apply. Label license, without restriction to scale, is granted to end-user upon purchase.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kevin W C Poon et al.
The Journal of organic chemistry, 71(10), 3923-3927 (2006-05-06)
2-Benzyloxy-1-methylpyridinium triflate (1) is a stable, neutral organic salt that converts alcohols into benzyl ethers upon warming. The synthesis and reactivity of 1 are described herein. Benzylation of a wide range of alcohols occurs in good to excellent yield.
Jumreang Tummatorn et al.
The Journal of organic chemistry, 72(23), 8962-8964 (2007-10-16)
Triethylamine (Et3N) mediates esterification reactions between the title reagent (1) and carboxylic acids. Alcohols, phenols, amides, and other sensitive functionality are not affected; a dual role for Et3N as a promoter and a scavenger is postulated. Benzyl esters are obtained
Dudley, G. B. et al.
Synlett, 3142-3142 (2005)

Articles

Dudley Reagents provide mild protection for alcohols and carboxylic acids in synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service