Skip to Content
Merck
All Photos(2)

Key Documents

679127

Sigma-Aldrich

7-Bromoisatin

96%

Synonym(s):

7-Bromoindole-1H-2,3-dione

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$86.80

$86.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$86.80

About This Item

Empirical Formula (Hill Notation):
C8H4BrNO2
CAS Number:
Molecular Weight:
226.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$86.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

mp

191-198 °C

functional group

bromo
ketone

SMILES string

Brc1cccc2C(=O)C(=O)Nc12

InChI

1S/C8H4BrNO2/c9-5-3-1-2-4-6(5)10-8(12)7(4)11/h1-3H,(H,10,11,12)

InChI key

OCVKSIWBTJCXPV-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Cale C Streeter et al.
Biochemistry, 58(17), 2260-2268 (2019-04-10)
The continued rise of antibiotic-resistant infections coupled with the limited pipeline of new antimicrobials highlights the pressing need for the development of new antibacterial agents. One potential pathway for new agents is de novo purine biosynthesis as studies have shown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service