Skip to Content
Merck
All Photos(1)

Key Documents

678538

Sigma-Aldrich

(−)-Ipc2B(allyl)borane solution

1 M in pentane

Synonym(s):

(-)-Ipc2B(allyl) solution, (−)-B-Allyldiisopinocampheylborane solution

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$411.00

$411.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$411.00

About This Item

Linear Formula:
(C10H17)2BCH2CH=CH2
CAS Number:
Molecular Weight:
326.37
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$411.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

optical activity

[α]20/D −35 to ±5°, c = 1

Quality Level

concentration

1 M in pentane

bp

35-36 °C

density

0.638 g/mL at 25 °C

storage temp.

−20°C

SMILES string

C[C@H]1[C@@H](C[C@@H]2C[C@H]1C2(C)C)B(CC=C)[C@@H]3C[C@@H]4C[C@H]([C@H]3C)C4(C)C

InChI

1S/C23H39B/c1-8-9-24(20-12-16-10-18(14(20)2)22(16,4)5)21-13-17-11-19(15(21)3)23(17,6)7/h8,14-21H,1,9-13H2,2-7H3/t14-,15-,16+,17+,18-,19-,20-,21-/m1/s1

InChI key

ZIXZBDJFGUIKJS-RLEROFIGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This allylboration reagent is a convenient, salt-free, stable solution of a chiral allyl borane for asymmetric allylation of aldehydes leading to chiral homoallylic alcohols.[1][2]

Other Notes

Material may become hazy upon cooler storage, this will not affect use
Under refrigerator storage, these salt-free, Ipc2B(allyl) reagents do not show any appreciable decrease in selectivity is observed after several months. They exhibit excellent reactivity and reactions can be performed at 5°C, even in water.[3]

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile route to 2, 6-dideoxyamino sugars from non-sugar materials: Syntheses of vicenisamine and kedarosamine.
Matsushima, Yoshitaka, et al.
J. Chem. Soc., Perkin Trans., 1.6, 569-577 (2001)
Catalysis?Based Total Synthesis of Latrunculin B
Furstner, Alois, et al.
Angewandte Chemie (International Edition in English), 115.43, 5516-5518 (2003)
Total synthesis of hypermodified epothilone analogs with potent in vitro antitumor activity
Kuzniewski, Christian N., et al.
Organic Letters, 10.6, 1183-1186 (2008)
Stereoselective Synthesis of 2, 6-syn-Dimethyl-tetrahydropyran Derivatives, Important Segments of Marine Polycyclic Ethers, by Unique Insertion of the Methyl Group
Maemoto, Michihiro, Atsushi Kimishima, and Tadashi Nakata.
Organic Letters, 11.21, 4814-4817 (2009)
Total synthesis of cryptomoscatone F1 through an asymmetric aldol approach
Ramesh, Perla, Atla Raju, and Nitin W. Fadnavis.
Tetrahedron Asymmetry, 26.21, 1251-1255 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service