Skip to Content
Merck
All Photos(1)

Key Documents

675679

Sigma-Aldrich

1-Methyl-2-(tributylstannyl)pyrrole

95%

Synonym(s):

(1-Methylpyrrol-2-yl)tributyltin, 2-(Tributylstannyl)-1-methylpyrrole, N-Methyl-2-(tributylstannyl)pyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H3C(C4H3N)]Sn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
370.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.5107

density

1.1223 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CCCC[Sn](CCCC)(CCCC)c1cccn1C

InChI

1S/C5H6N.3C4H9.Sn/c1-6-4-2-3-5-6;3*1-3-4-2;/h2-4H,1H3;3*1,3-4H2,2H3;

InChI key

DINAKCGOEKXDTP-UHFFFAOYSA-N

Application

1-Methyl-2-(tributylstannyl)pyrrole can be used as a reactant to prepare:
  • Bithienylpyrrole derivatives through a Stille coupling reaction with bithienyl bromides in the presence of Pd catalyst.
  • Di(bisthienyl)-o-carborane (DBTC) by reacting with 5,5′-dibromo-o-carboranyl-bisthiophenes using Pd catalyst. DBTC is electropolymerized to produce conducting polymers.
  • Poly(Py-BTz-Py) (2-butyl-4,7-bis(1-methyl-1H-pyrrol-2-yl)-2H-benzo[d][1,2,3]triazole) via Stille coupling reaction. Poly(Py-BTz-Py) can be used as an organic photocatalyst for the formation C-C bond under visible light irradiation.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

208.9 °F - closed cup

Flash Point(C)

98.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Design of Donor-Acceptor-Type Organic Photocatalysts for Metal-free Aromatic C- C Bond Formations under Visible Light
Wang L, et al.
advanced synthesis and catalysis, 360(22), 4312- 4318 (2018)
Synthesis of π-conjugated systems bearing thiophene and pyrrole heterocycles through palladium catalyzed cross-coupling reactions
Castro MCR and Raposo MMM
Tetrahedron, 72(15), 1881-1887 (2016)
Molecular Design of Donor-Acceptor-Type Organic Photocatalysts for Metal-free Aromatic C- C Bond Formations under Visible Light
Wang L, et al.
Advanced Synthesis & Catalysis, 360(22), 4312- 4318 (2018)
Synthesis and electropolymerization of a series of 2, 2?-(ortho-carboranyl) bisthiophenes
Zhao N, et al.
Journal of Organometallic Chemistry, 828(15), 157- 165 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service