Skip to Content
Merck
All Photos(1)

Key Documents

675563

Sigma-Aldrich

1,2-Bis[(2S,5S)-2,5-dimethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40F3O3P2RhS
CAS Number:
Molecular Weight:
666.52
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

136.0-154.7 °C

SMILES string

[Rh+].[O-]S(=O)(=O)C(F)(F)F.C1CC=CCCC=C1.C[C@H]2CC[C@H](C)P2c3ccccc3P4[C@@H](C)CC[C@@H]4C

InChI

1S/C18H28P2.C8H12.CHF3O3S.Rh/c1-13-9-10-14(2)19(13)17-7-5-6-8-18(17)20-15(3)11-12-16(20)4;1-2-4-6-8-7-5-3-1;2-1(3,4)8(5,6)7;/h5-8,13-16H,9-12H2,1-4H3;1-2,7-8H,3-6H2;(H,5,6,7);/q;;;+1/p-1/b;2-1-,8-7-;;/t13-,14-,15-,16-;;;/m0.../s1

InChI key

AKJDLXZYNLUJQJ-KZOFNLLLSA-M

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Asymmetric hydrogenation enables scalable synthesis of single-enantiomer compounds with minimal byproducts, ideal for commercial manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service