Skip to Content
Merck
All Photos(1)

Key Documents

672688

Sigma-Aldrich

O-(2-Carboxyethyl)-O′-(2-mercaptoethyl)heptaethylene glycol

≥95% (oligomer purity)

Synonym(s):

O-(2-Mercaptoethyl)-O′-(2-carboxyethy)heptaethylene glycol, PEG thiol acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$528.00

$528.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$528.00

About This Item

Empirical Formula (Hill Notation):
C19H38O10S
CAS Number:
Molecular Weight:
458.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$528.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

≥95% (oligomer purity)

reaction suitability

reagent type: cross-linking reagent

storage temp.

−20°C

SMILES string

OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCS

InChI

1S/C19H38O10S/c20-19(21)1-2-22-3-4-23-5-6-24-7-8-25-9-10-26-11-12-27-13-14-28-15-16-29-17-18-30/h30H,1-18H2,(H,20,21)

InChI key

SKYJRDAORYSCAL-UHFFFAOYSA-N

Application

O-(2-Carboxyethyl)-O′-(2-mercaptoethyl)heptaethylene glycol can be used:
  • To prepare near-infrared fluorochromes applicable in imaging techniques.[1]
  • To functionalize Au nanoparticles for optoelectronic applications.[2]
  • As a linker in the synthesis of antibody-functionalized Au nanoparticles for forensic applications.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monofunctional near-infrared fluorochromes for imaging applications
Hilderbrand SA, et al.
Bioconjugate Chemistry, 16(5), 1275-1281 (2005)
Evaluation of multi-target immunogenic reagents for the detection of latent and body fluid-contaminated fingermarks
Lam R, et al.
Forensic Science International, 264(5), 168-175 (2016)
Scott A Hilderbrand et al.
Bioconjugate chemistry, 16(5), 1275-1281 (2005-09-22)
In this report, the development of a new class of monocarboxylate functionalized cyanine derivatives using improved synthetic procedures is detailed. The employed synthetic strategy relies on efficient nucleophilic attack of alkyl-thiols on cyanine dyes bearing chloro-substituted polymethinic linkers. Monocarboxylate derivatized
Controlled assembly of plasmonic nanoparticles using neutral-charged diblock copolymers
Yin Q, et al.
Journal of Colloid and Interface Science, 431(5), 97-104 (2014)
Primiano Pio Di Mauro et al.
International journal of pharmaceutics, 553(1-2), 169-185 (2018-10-16)
A dual-targeting drug delivery system for paclitaxel (PTX) was developed by functionalizing novel polyester-based nanoparticles (NPs) with peptides possessing special affinity for low-density lipoprotein receptor (LDLR), overcoming the limitations of the current chemotherapeutics, to transport drug from blood to brain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service