Skip to Content
Merck
All Photos(1)

Key Documents

672297

Sigma-Aldrich

(S)-N,N′-Dimethyl-1,1′-binaphthyldiamine

≥99.0%

Synonym(s):

(S)-N,N′-Dimethyl-1,1′-binaphthalene-2,2′--diamine, (S)-N,N′-Dimethyl-2,2′-diamino-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$117.00

$117.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$117.00

About This Item

Empirical Formula (Hill Notation):
C22H20N2
CAS Number:
Molecular Weight:
312.41
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$117.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Assay

≥99.0% (HPLC)
≥99.0%

optical purity

enantiomeric excess: ≥99.0% (HPLC)

functional group

amine

storage temp.

2-8°C

SMILES string

CNc1ccc2ccccc2c1-c3c(NC)ccc4ccccc34

InChI

1S/C22H20N2/c1-23-19-13-11-15-7-3-5-9-17(15)21(19)22-18-10-6-4-8-16(18)12-14-20(22)24-2/h3-14,23-24H,1-2H3

InChI key

PXDKDUXMEHPNCO-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service