Skip to Content
Merck
All Photos(1)

Key Documents

672017

Sigma-Aldrich

2-(Dicyclohexylphosphino)-1-phenyl-1H-pyrrole

95%

Synonym(s):

cataCXium® PCy

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$290.00

$290.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$290.00

About This Item

Empirical Formula (Hill Notation):
C22H30NP
CAS Number:
Molecular Weight:
339.45
Beilstein:
9649724
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$290.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Quality Level

Assay

95%

reaction suitability

reaction type: Asymmetric synthesis
reagent type: ligand
reaction type: Aminations

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

functional group

phosphine

SMILES string

C1CCC(CC1)P(C2CCCCC2)c3cccn3-c4ccccc4

InChI

1S/C22H30NP/c1-4-11-19(12-5-1)23-18-10-17-22(23)24(20-13-6-2-7-14-20)21-15-8-3-9-16-21/h1,4-5,10-12,17-18,20-21H,2-3,6-9,13-16H2

InChI key

RVFZOTGUPNHVRA-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Legal Information

WO 2004/101581
cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service