Skip to Content
Merck
All Photos(2)

Key Documents

668990

Sigma-Aldrich

Tris(diethylamido)(tert-butylimido)tantalum(V)

packaged for use in deposition systems

Synonym(s):

TBTDET, Tris(diethylamino)(tert-butylimino)tantalum(V)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$3,200.00

$3,200.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$3,200.00

About This Item

Linear Formula:
(CH3)3CNTa(N(C2H5)2)3
CAS Number:
Molecular Weight:
468.46
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$3,200.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.99% (trace metals analysis)

form

liquid

reaction suitability

core: tantalum

density

1.252 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)[Ta](=NC(C)(C)C)(N(CC)CC)N(CC)CC

InChI

1S/C4H9N.3C4H10N.Ta/c1-4(2,3)5;3*1-3-5-4-2;/h1-3H3;3*3-4H2,1-2H3;/q;3*-1;+3

InChI key

YYKBKTFUORICGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 73 Tantalum

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service