Skip to Content
Merck
All Photos(2)

Key Documents

668729

Sigma-Aldrich

Diethylzinc

packaged for use in deposition systems

Synonym(s):

DEZn, Et2Zn, DEZ, Zincdiethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2Zn
CAS Number:
Molecular Weight:
123.51
Beilstein:
3587207
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

117 °C (lit.)

mp

−28 °C (lit.)

density

1.205 g/mL at 25 °C (lit.)

SMILES string

CC[Zn]CC

InChI

1S/2C2H5.Zn/c2*1-2;/h2*1H2,2H3;

InChI key

HQWPLXHWEZZGKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 30 Zinc

Application

Reacted with water in a low temperature (<200°C) atomic layer deposition of polycrystalline ZnO layers displaying exciton photoluminescence at room temperature.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Applied Physics, 103, 033515-033515 (2008)
Zhiyan Huang et al.
The Journal of organic chemistry, 72(4), 1373-1378 (2007-02-10)
A novel class of sulfinamido alcohol ligands 1-6 was synthesized from (S)-tert-butanesulfinamide. These ligands showed excellent catalytic activities and enantiomeric selectivities in asymmetric additions of diethylzinc to aromatic aldehydes.
Efficient chirality switching in the addition of diethylzinc to aldehydes in the presence of simple chiral alpha-amino amides.
M Isabel Burguete et al.
Angewandte Chemie (International ed. in English), 46(47), 9002-9005 (2007-09-26)
Isabelle Bonnaventure et al.
The Journal of organic chemistry, 73(16), 6330-6340 (2008-07-22)
The hemilabile ligand Me-DuPHOS(O) 2 has proven to be a successful ligand for the copper-catalyzed addition of diethylzinc to N-phosphinoylimines. The corresponding alpha-chiral amines were obtained in high yields (80-98%) and enantiomeric ratios (19.0:1 to 99.0:1 er). Furthermore, this Cu*
Weimin Lin et al.
The Journal of organic chemistry, 74(2), 645-651 (2008-12-06)
The application of a zinc carbenoid-mediated chain-extension reaction to a functionalized peptide isostere is reported. The cleavage site of human CVM protease was utilized as a target for testing the synthetic methodology. The utility of this chain-extension reaction is demonstrated

Articles

Atomic layer deposition meets various needs including semiconductor device miniaturization and nanoparticle coating.

Atomic Layer Deposition (ALD) technology ensures uniform coating on complex 3D surfaces with precise chemisorption cycles.

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

See All

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service