Skip to Content
Merck
All Photos(1)

Documents

668516

Sigma-Aldrich

(S)-(+)-3,3-Dimethyl-2-butylamine

97%

Synonym(s):

(S)-3,3-Dimethyl-2-aminobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)3C)CH(NH2)CH3
CAS Number:
Molecular Weight:
101.19
Beilstein:
4652328
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.413

bp

103 °C

mp

-4 °C

density

0.743 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C[C@H](N)C(C)(C)C

InChI

1S/C6H15N/c1-5(7)6(2,3)4/h5H,7H2,1-4H3/t5-/m0/s1

InChI key

DXSUORGKJZADET-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-3,3-Dimethyl-2-butylamine may be used in the preparation of:
  • methyl 3′-{[(2S-3,3-dimethylbutan-2-yl]carbamoyl]-2,2′-binaphthalene-3-carboxylate
  • N,N-(S)-bis(diphenylphosphanyl)-3,3-dimethyl-2-butylamine

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Induced circular dichroism in chiral N-methyl amides possessing an achiral binaphthyl chromophore and its application to absolute configuration determination of aliphatic chiral amines.
Fujiwara T, et al.
Tetrahedron Asymmetry, 23(13), 981-991 (2012)
Efficient Synthesis of Novel N-Substituted Bulky Diphosphinoamines.
Maumela MC, et al.
Synthesis, 2007(24), 3863-3867 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service