668206
(R)-(−)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)dimethylamine
97%
Synonym(s):
(R)-MonoPhos®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C22H18NO2P
CAS Number:
Molecular Weight:
359.36
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
190 °C (dec.)
functional group
amine
SMILES string
CN(C)P1Oc2ccc3ccccc3c2-c4c(O1)ccc5ccccc45
InChI
1S/C22H18NO2P/c1-23(2)26-24-19-13-11-15-7-3-5-9-17(15)21(19)22-18-10-6-4-8-16(18)12-14-20(22)25-26/h3-14H,1-2H3
InChI key
QCHAVHXSBZARBO-UHFFFAOYSA-N
Legal Information
Sold under license from DSM for research purposes only.
MonoPhos is a registered trademark of DSM IP Assets B.V.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service