Skip to Content
Merck
All Photos(2)

Key Documents

667366

Sigma-Aldrich

6-Bromo-2-chlorobenzothiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3BrClNS
CAS Number:
Molecular Weight:
248.53
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

99-103 °C

functional group

bromo
chloro

SMILES string

Clc1nc2ccc(Br)cc2s1

InChI

1S/C7H3BrClNS/c8-4-1-2-5-6(3-4)11-7(9)10-5/h1-3H

InChI key

IJQSMNIZBBEBKI-UHFFFAOYSA-N

Application

Starting material in a synthesis of benzothiazole dimers with high binding affinity to β-amyloid fibrils.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong Rim Byeon et al.
Bioorganic & medicinal chemistry letters, 17(14), 4022-4025 (2007-06-05)
New ferulic acid and benzothiazole dimer derivatives were synthesized and evaluated by in vitro competition assay using [(125)I]TZDM for their specific binding affinities to Abeta fibrils. In particular, 4a showed the most excellent binding affinity (K(i)=0.53 nM), compared to PIB

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service