Skip to Content
Merck
All Photos(1)

Key Documents

666459

Sigma-Aldrich

6-Indolylboronic acid

≥95%

Synonym(s):

6-Indoleboronic acid, Indole-6-boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$134.00

$134.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$134.00

About This Item

Linear Formula:
(C8H6N)B(OH)2
CAS Number:
Molecular Weight:
160.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$134.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95%

form

powder

mp

177-181 °C

storage temp.

2-8°C

SMILES string

OB(O)c1ccc2cc[nH]c2c1

InChI

1S/C8H8BNO2/c11-9(12)7-2-1-6-3-4-10-8(6)5-7/h1-5,10-12H

InChI key

ZVMHOIWRCCZGPZ-UHFFFAOYSA-N

Application

Reactant involved in synthesis of:
  • Indole compounds for use as HIV-1 glycoprotein-41 fusion inhibitors
  • δ-Carbolines / carbozoles
  • Trisubstituted pyrimidines as PI3K inhibitors
  • (Thienopyridine)carboxamides as CHK1 inhibitors
  • cis-Fluorostilbenes

Reactant involved in Suzuki-Miyaura reactions
Used in a study of racemization in Suzuki cross-couplings of arylamino acids.[1]

Other Notes

May contain varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica Prieto et al.
The Journal of organic chemistry, 72(3), 1047-1050 (2007-01-27)
Reaction conditions considered to be typical in Suzuki couplings can cause significant (up to 34% of the unwanted enantiomer) loss of optical purity in sensitive substrates such as hydroxyphenylglycine 1. This may be remedied using sodium succinate instead of sodium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service