666270
Methyl 3-amino-2-(trifluoromethyl)propionate
90%
Synonym(s):
Methyl 2-(aminomethyl)-3,3,3-trifluoro-propanoate
About This Item
Recommended Products
Assay
90%
form
liquid
refractive index
n20/D 1.374
bp
50 °C/0.2 mmHg
density
1.345 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
COC(=O)C(CN)C(F)(F)F
InChI
1S/C5H8F3NO2/c1-11-4(10)3(2-9)5(6,7)8/h3H,2,9H2,1H3
InChI key
IWNHSPZURKMJSI-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
213.8 °F - closed cup
Flash Point(C)
101 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service