Skip to Content
Merck
All Photos(1)

Key Documents

666122

Sigma-Aldrich

α,α,α′,α′-Tetramethyl-1,3-benzenedipropionic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$77.10

$77.10


Estimated to ship onApril 27, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$77.10

About This Item

Empirical Formula (Hill Notation):
C16H22O4
CAS Number:
Molecular Weight:
278.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$77.10


Estimated to ship onApril 27, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

132-136 °C

functional group

carboxylic acid

SMILES string

CC(C)(Cc1cccc(CC(C)(C)C(O)=O)c1)C(O)=O

InChI

1S/C16H22O4/c1-15(2,13(17)18)9-11-6-5-7-12(8-11)10-16(3,4)14(19)20/h5-8H,9-10H2,1-4H3,(H,17,18)(H,19,20)

InChI key

BZWQVGTVYGEXTE-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Chu Chen et al.
Genes & cancer, 7(5-6), 148-153 (2016-08-24)
Protein kinases are attractive drug targets for numerous human diseases including cancers, diabetes and neurodegeneration. A number of kinase inhibitors that covalently target a cysteine residue in their target kinases have recently entered use in the cancer clinic. Despite the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service