Skip to Content
Merck
All Photos(1)

Documents

665150

Sigma-Aldrich

Ethyl N-Boc-piperidine-4-carboxylate

97%

Synonym(s):

Ethyl 1-(tert-Butyloxycarbonyl)isonipecotate, Ethyl 1-(tert-butoxycarbonyl)-4-piperidinecarboxylate, Piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23NO4
CAS Number:
Molecular Weight:
257.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.458

bp

120-135 °C/0.5 mmHg

density

1.046 g/mL at 25 °C

SMILES string

CCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C

InChI

1S/C13H23NO4/c1-5-17-11(15)10-6-8-14(9-7-10)12(16)18-13(2,3)4/h10H,5-9H2,1-4H3

InChI key

MYHJCTUTPIKNAT-UHFFFAOYSA-N

Application

Substrate used in a palladium-catalyzed α-arylation of esters with heterocyclic bromides and chlorides leading to, for example, 4-pyridylpiperidinyl esters.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 48, 1191-1191 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service