Skip to Content
Merck
All Photos(1)

Key Documents

665010

Sigma-Aldrich

2,2′-Bis[(S-1,1′-binaphthyl-2,2′-dimethyl)phosphino]diethylamine

97%

Synonym(s):

Bis(S-2-(4,5-dihydro-3H-binaphtho[1,2-c;2′,1′-e]-phosphepino)ethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H41NP2
Molecular Weight:
693.79
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

230-236 °C

functional group

amine
phosphine

SMILES string

C(CP1Cc2ccc3ccccc3c2-c4c(C1)ccc5ccccc45)NCCP6Cc7ccc8ccccc8c7-c9c(C6)ccc%10ccccc9%10

InChI

1S/C48H41NP2/c1-5-13-41-33(9-1)17-21-37-29-50(30-38-22-18-34-10-2-6-14-42(34)46(38)45(37)41)27-25-49-26-28-51-31-39-23-19-35-11-3-7-15-43(35)47(39)48-40(32-51)24-20-36-12-4-8-16-44(36)48/h1-24,49H,25-32H2

InChI key

VMLOJVKKOABMEO-UHFFFAOYSA-N

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service