Skip to Content
Merck
All Photos(2)

Key Documents

663107

Sigma-Aldrich

(2S,5S)-(−)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone

97%

Synonym(s):

(2S,5S)-2-tert-Butyl-3-methyl-5-phenylmethyl-4-imidazolidinone, (2S,5S)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O
CAS Number:
Molecular Weight:
246.35
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

93-100 °C (lit.)

functional group

phenyl

SMILES string

CN1[C@H](N[C@@H](Cc2ccccc2)C1=O)C(C)(C)C

InChI

1S/C15H22N2O/c1-15(2,3)14-16-12(13(18)17(14)4)10-11-8-6-5-7-9-11/h5-9,12,14,16H,10H2,1-4H3/t12-,14-/m0/s1

InChI key

SKHPYKHVYFTIOI-JSGCOSHPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone is a chiral imidazolidinone organocatalyst, developed by MacMillan and co-workers.[1]

Application

(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone is a second-generation MacMillan catalyst, which can be used as a chiral organocatalyst in:
  • The chiral transformation reaction, including Friedel-Crafts and Mukaiyama-Michael reactions.[2]
  • The preparation of substituted spiroundecenetriones via asymmetric domino Knoevenagel/Diels-Alder reactions.[3]
  • The asymmetric synthesis of β-hydroxy aldehydes and their dimethylacetals via aldehyde-aldehyde aldol condensation reaction.[4]
  • The enantioselective α-fluorination of aldehydes using N-fluorobenzenesulfonamide as a fluorinating agent.[5]
  • The stereoselective preparation of (oxomethyl)oxabicyclo[3.2.1]octenones and tricyclic pyrroles via [4+3] cycloaddition of (trialkylsiloxy)pentadienals to furans.[6]

Metal-free OrganoCatalyst technology for asymmetric catalysis. Catalyzes asymmetric indole alkylations, Friedel-Crafts alkylations, and a broad range of conjugate addition reactions in high enantiomeric excess.

Features and Benefits

Advantages of MacMillan imidazolidinone organocatalysts:[1]
  • Superior enantiocontrol in numerous transformations
  • High activities at low catalyst loadings
  • Extraordinary functional group tolerance

Legal Information

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa D Beeson et al.
Science (New York, N.Y.), 316(5824), 582-585 (2007-03-31)
The asymmetric alpha-addition of relatively nonpolar hydrocarbon substrates, such as allyl and aryl groups, to aldehydes and ketones remains a largely unsolved problem in organic synthesis, despite the wide potential utility of direct routes to such products. We reasoned that
The Importance of Iminium Geometry Control in Enamine Catalysis: Identification of a New Catalyst Architecture for Aldehyde-Aldehyde Couplings
Mangion IK, et al.
Angewandte Chemie (International Edition in English), 116(48), 6890-6892 (2004)
Asymmetric organocatalysis of 4+ 3 cycloaddition reactions
Harmata M, et al.
Journal of the American Chemical Society, 125(8), 2058-2059 (2003)
Organocatalytic Asymmetric Domino Knoevenagel/Diels-Alder Reactions: A Bioorganic Approach to the Diastereospecific and Enantioselective Construction of Highly Substituted Spiro [5, 5] undecane-1, 5, 9-triones
Ramachary DB, et al.
Angewandte Chemie (International Edition in English), 42(35), 4233-4237 (2003)
Nick A Paras et al.
Journal of the American Chemical Society, 124(27), 7894-7895 (2002-07-04)
The first enantioselective organocatalytic alkylation of electron-rich benzene rings with alpha,beta-unsaturated aldehydes has been accomplished. The use of iminium catalysis has provided a new strategy for the enantioselective construction of benzylic stereogenicity, an important chiral synthon for natural product and

Articles

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service