Skip to Content
Merck
All Photos(1)

Key Documents

660450

Sigma-Aldrich

3-Aminopropyl(3-oxobutanoic acid) functionalized silica

nanoparticles, ~15 nm particle size (DLS), dispersion, 2.5 % (w/v) in DMF

Synonym(s):

Carboxylic acid-functionalized silica colloid

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$1,220.00

$1,220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$1,220.00

About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$1,220.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

dispersion

concentration

2.5 % (w/v) in DMF

particle size

~15 nm (DLS)

bp

143-153 °C

density

0.927 g/mL at 25 °C

functional group

carboxylic acid

storage temp.

2-8°C

Application

Dispersion may settle. Shake well before use.
Used for the self-assembly of nano-scale monolayers.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

90.0 °F - closed cup

Flash Point(C)

32.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Galow, T.H. et al.
Advanced Materials, 12, 576-576 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service