Skip to Content
Merck
All Photos(1)

Key Documents

65980

Sigma-Aldrich

(S)-(−)-2-Methylbutanol

≥95.0% (sum of enantiomers, GC)

Synonym(s):

(S)-(−)-2-Methyl-1-butanol, Active amyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$121.00
500 ML
$513.00

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$121.00
500 ML
$513.00

About This Item

Empirical Formula (Hill Notation):
C5H12O
CAS Number:
Molecular Weight:
88.15
Beilstein:
1718809
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95.0% (sum of enantiomers, GC)

optical activity

[α]20/D −6.3±0.5°, c = 10% in ethanol

refractive index

n/D 1.41
n20/D 1.411

bp

129-131 °C (lit.)

density

0.819 g/mL at 20 °C (lit.)
0.811 g/mL at 25 °C

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

CC[C@H](C)CO

InChI

1S/C5H12O/c1-3-5(2)4-6/h5-6H,3-4H2,1-2H3/t5-/m0/s1

InChI key

QPRQEDXDYOZYLA-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

rest mainly 3-methyl-1-butanol

Application

(S)-(-)-2-Methylbutanol can be used as a reactant to prepare:
  • Methylbutyl-2-(3-thienyl)acetate (MBTA) by esterification reaction with 3-thiophene acetic acid in the presence of an acid catalyst.[1]
  • (2S)-2-Methyl-1-butanesulfenyl chloride by reacting with thioacetic acid using Mitsunobu reaction conditions.[2]
  • (+)-Violapyrone C and (-)-myxalamide A.[3][4]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

108.5 °F

Flash Point(C)

42.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total Synthesis and Configurational Validation of (+)-Violapyrone C
Lee JS, et al.
European Journal of Organic Chemistry, 2014(21), 4472-4476 (2014)
Synthesis and Characterization of Conducting Copolymers of (S)-2-Methylbutyl-2-(3-thienyl) acetate with Pyrrole and Thiophene
Levent A, et al.
Macromolecular Chemistry and Physics, 204(8), 1118-1122 (2003)
A general method for the enantiospecific synthesis of optically active aliphatic sulfenyl chlorides and thiophthalimides
Cevasco G, et al.
Tetrahedron Asymmetry, 1(3), 141-142 (1990)
Concise Total Synthesis of (-)-Myxalamide A
Fujita K et al.
Angewandte Chemie (International ed. in English), 51(29), 7271-7274 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service