Skip to Content
Merck
All Photos(1)

Key Documents

659533

Sigma-Aldrich

2-Amino-2-methyl-1-propanol-d11

97 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2C(ND2)CD2OD
CAS Number:
Molecular Weight:
100.20
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

97 atom % D

Assay

98% (CP)

form

solid

bp

165 °C

mp

24-28 °C

density

1.048 g/mL at 25 °C

mass shift

M+11

SMILES string

[2H]OC([2H])([2H])C(N([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3/i1D3,2D3,3D2,6D/hD2

InChI key

CBTVGIZVANVGBH-BIGNBOPISA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

154.0 °F - closed cup

Flash Point(C)

67.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service