Skip to Content
Merck
All Photos(1)

Documents

65700

Sigma-Aldrich

N6-Methyladenine

≥99.0% (HPLC)

Synonym(s):

6-(Methylamino)purine, N-Methyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N5
CAS Number:
Molecular Weight:
149.15
Beilstein:
7707
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99.0% (HPLC)

mp

≥300 °C

SMILES string

CNc1ncnc2[nH]cnc12

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service