Skip to Content
Merck
All Photos(1)

Key Documents

65659

Sigma-Aldrich

12-(Methylamino)dodecanoic acid

≥98.0% (NT)

Synonym(s):

12-(Methylamino)lauric acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$468.00

$468.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$468.00

About This Item

Linear Formula:
CH3NH(CH2)11COOH
CAS Number:
Molecular Weight:
229.36
Beilstein:
6123408
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$468.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (NT)

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

139-143 °C

solubility

methanol: 50 mg/mL, clear

application(s)

peptide synthesis

SMILES string

CNCCCCCCCCCCCC(O)=O

InChI

1S/C13H27NO2/c1-14-12-10-8-6-4-2-3-5-7-9-11-13(15)16/h14H,2-12H2,1H3,(H,15,16)

InChI key

XTXRYZLZPWMJBM-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service