Recommended Products
Assay
97%
refractive index
n20/D 1.5707 (lit.)
bp
110-112 °C/10 mmHg
density
1.4569 g/mL at 25 °C (lit.)
functional group
bromo
chloro
SMILES string
Clc1ccccc1CCBr
InChI
1S/C8H8BrCl/c9-6-5-7-3-1-2-4-8(7)10/h1-4H,5-6H2
InChI key
AECBVDLERUETKG-UHFFFAOYSA-N
1 of 4
This Item | 205486 | P4758 | B0126 |
---|---|---|---|
practical grade | indicator grade | - | - |
ethanol: 50 mg/mL, clear, red-orange | - | water: 1 mg/mL, opaque, dark red | methanol: 10 mg/mL |
diagnostic assay manufacturing | diagnostic assay manufacturing | diagnostic assay manufacturing | diagnostic assay manufacturing |
powder | powder | powder | powder |
>80% NaOH basis (titration) | - | - | - |
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service